Learn More
Cayman Chemical 1 2DIpalmItoyl3Oleoylrac 10mg
SDP
Supplier:  Cayman Chemical 2687810 MG
Encompass_Preferred
A triacylglycerol; has been found in Apiaceae seed oils, lard, tallow, as well as palm oil, olein, and stearin; has been used as a substrate to determine the regioselectivity and substrate specificity of lipases from Geotrichum species
Catalog No. 50-293-4877