Learn More
Cayman Chemical 1 2DIpentadecnoylracglyce 10mg
SDP
Supplier:  Cayman Chemical 2694110 MG
Encompass_Preferred
A diacylglycerol; has been used as an internal standard for the quantification of 1,2-diacylglycerols and component fatty acids in rat myocardial tissue by capillary gas chromatography
Catalog No. 50-293-7634