Learn More
Cayman Chemical 1 2DIstearoyl3Decnoylrac 10mg
SDP
Supplier:  Cayman Chemical 2683110 MG
Encompass_Preferred
A triacylglycerol; contains stearic acid at the sn-1 and sn-2 positions and decanoic acid at the sn-3 position
Catalog No. 50-293-2919