Learn More
Cayman Chemical 1 2DIstearoyld70snglycero 5mg
SDP
Supplier:  Cayman Chemical 287525 MG
Encompass_Preferred
An internal standard for the quantification of 1,2-distearoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-294-0324