Learn More
Andwin Scientific 1/2" Laser Tough-Spots, Green, 3840/pk
SDP
Supplier:  Andwin Scientific SPOT2200
Encompass_Preferred
Tough-Spots are chemically inert and flexible white and colored polyolefin labels that adhere to most plastics (best performance with virgin polypropylene) as well as glass and metals.
Catalog No. NC1660221