Learn More
Accuflow Systems Saint-Gobain Sani-Tech Silicone Tubing, Platinum-Cured, 1/2 in ID, 25 ft
Supplier:  Accuflow Systems STHTC5004
Encompass_Preferred
Platinum-cured silicone tubing with an ultra-pure inner bore for biopharmaceutical and laboratory fluid transfer. Designed for repeated autoclaving and single-use assemblies, it delivers consistent flow characteristics and low extractables.
  • Platinum-cured silicone material suitable for pharmaceutical use.
  • Inner diameter 1/2 in, outer diameter 3/4 in, wall thickness 1/8 in.
  • Length 25 ft coil for assembly or replacement use.
  • Temperature range -80°F (-62°C) to 500°F (260°C); autoclavable.
  • Low extractables and smooth inner bore to minimize product adhesion.
  • Laser-marked traceability available for sizes 1/16 in and greater.
Catalog No. NC2312824