Learn More
Grainger WESTWARD Garden Hose Adapter, 1/2 in
Supplier:  Grainger 4KG83
Encompass_Preferred
The WESTWARD Garden Hose Adapter is a 1/2 inch brass hose adapter that connects a garden hose to a water outlet or faucet. It has standard garden hose threads on one end and a male pipe thread on the other end. The brass construction provides durability. This adapter allows you to easily connect a garden hose to an outdoor faucet or spigot. It is made in the USA by Westward, a manufacturer of high quality plumbing tools and accessories. This is a useful adapter for connecting hoses in your garden or outdoor space.
Catalog No. NC2754332