Learn More
Cayman Chemical 1 2ODIoctadecylracglycRo 500mg
SDP
Supplier:  Cayman Chemical 16491500 MG
Encompass_Preferred
A saturated dialkyl glyceryl ether with octadecyl groups at the sn-1 and sn-2 positions, which mimics the structure of diacylglycerol
Catalog No. 50-297-0483