Learn More
Medchemexpress LLC 1-[(2R)-1-methylsulfonylpropan-2-yl]-4-(trifluoromethyl)indole-5-carbonitrile | 1539314-06-1 | MFCD30343844 | 99.4% | 330.33 g/mol | C14H13F3N2O2S | 10 MG

Supplier: Medchemexpress LLC HY10018610MG
GSK-2881078 is a nonsteroidal selective androgen receptor modulator (SARM) supplied as a high-purity research compound for preclinical pharmacology and biochemical assays. The compound is provided as a powder suitable for controlled storage and use as a reference standard in laboratory studies.
- Orally active nonsteroidal selective androgen receptor modulator used in muscle wasting research.
- High purity research-grade powder (99.4%).
- Molecular formula C14H13F3N2O2S; molecular weight 330.33 g/mol.
- CAS number 1539314-06-1 for unambiguous identification.
- Recommended storage: powder at -20°C for long-term, 4°C for short-term.
- Supplied in 10 mg quantity suitable for small-scale assays.
- Suitable as a reference standard in pharmacology and ADME studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.