Learn More
Medchemexpress LLC 1-((2S,5S)-5-Ethynyl-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione | 634907-30-5 | 98.1% | 248.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16776A
Encompass_Preferred
(2S,5S)-Censavudine is the (2S,5S)-enantiomer of Censavudine, a nucleoside reverse transcriptase inhibitor and a potent HIV inhibitor. It also functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent HIV inhibitor
  • Nucleoside reverse transcriptase inhibitor
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
Catalog No. 50-003-95294