Learn More
Medchemexpress LLC 1,3,4,6-tetrachloro-3α,6α-diphenylglycouril | 51592-06-4 | MFCD00027367 | 98.0% | 432.09 g/mol | C16H10Cl4N4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW3420925MG
Encompass_Preferred
1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril is a small-molecule research reagent used primarily as an iodination reagent and oxidative agent for protein labeling and biochemical assays. It is supplied as a powder for laboratory use with documented purity and storage recommendations.
  • High purity suitable for research applications.
  • Suitable for in vitro iodination and protein labeling.
  • Available in small-mass vials for convenient handling.
  • Powder form with documented storage temperature ranges for stability.
  • Characterized molecular weight and chemical formula for experimental planning.
Catalog No. 50-002-80481