Learn More
Medchemexpress LLC 1-(3,4-dihydroxy-5-nitrophenyl)-2-phenylethanone | 274925-86-9 | MFCD09954136 | 99.9% | 273.24 | C14H11NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106405100MG
Encompass_Preferred
Nebicapone is a reversible catechol-O-methyltransferase (COMT) inhibitor provided as a high-purity research reagent for biochemical and pharmacological studies. It is characterized by known identifiers and physical properties, and is intended for laboratory research use only (not for human use).
  • Reversible COMT inhibitor suitable for enzyme assays and pharmacology studies.
  • High purity for analytical and experimental consistency.
  • Well-characterized identifiers available (CAS, MFCD, InChIKey).
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Supplied in a measured 100 mg amount for small-scale experiments.
Catalog No. 50-002-44031