Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3,4-oxadiazole | 288-99-3 | MFCD00955535 | 99.8% | 70.05 g/mol | C2H2N2O | 5 G
SDP

Supplier:  Medchemexpress LLC HYW1311225G

Encompass_Preferred

1,3,4-Oxadiazole is a small heterocyclic organic compound used as a research reagent and synthetic intermediate in medicinal chemistry. It is intended for laboratory research use only and is commonly employed in synthesis and structure-activity relationship studies of oxadiazole derivatives.

  • Small heterocyclic organic compound for research use.
  • Molecular formula C2H2N2O; molecular weight 70.05 g/mol.
  • High purity, typically 99.8%.
  • CAS number 288-99-3 for unambiguous identification.
  • Available in multiple package sizes, including 5 g.
  • Datasheet, certificate of analysis, and safety data sheet available.

Catalog No. 50-002-82845


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.