Learn More
Medchemexpress LLC 1-[(3,5-dichlorophenyl)sulfonyl]-N-[(4-fluorophenyl)methyl]-4-piperidinecarboxamide | 2227549-00-8 | 99.3% | 445.34 g/mol | C19H19Cl2FN2O3S | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12473810MM 1ML
Encompass_Preferred
BI01383298 is a small-molecule chemical probe that potently and selectively inhibits the human sodium-citrate co-transporter SLC13A5 (NaCT). It is supplied as a 10 mM solution in DMSO for in vitro research and shows low-nanomolar potency (IC50 ≈56 nM in HEK cells overexpressing SLC13A5; 24 nM in HepG2 cells) with >1,000-fold selectivity versus related family members. Typical supplied purity is ~99.3%, and the molecular formula is C19H19Cl2FN2O3S (MW ≈445 g/mol).
  • Potent SLC13A5 inhibition at low nanomolar concentrations.
  • High selectivity versus related transporters.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity suitable for biochemical assays.
  • Intended for in vitro research use; not recommended for mouse models.
Catalog No. 50-002-39404