Learn More
Medchemexpress LLC 1,3,5-triazine-1,3,5(2H,4H,6H)-triethanol | 4719-04-4 | MFCD01678788 | >96.0% | 219.28 g/mol | C9H21N3O3 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0181561KG
Encompass_Preferred
Triazinetriethanol (CAS 4719-04-4) is a triazine-based, volatile compound used in chemical research and reported as a component in some internal can coatings. It is supplied as a colorless to light yellow liquid with molecular weight 219.28 g/mol. The material is intended for research use only and should be handled and stored according to safety data guidance.
Catalog No. 50-003-00638