Learn More
Cayman Chemical 1 3 6 8PyrentetrasulfonIc A 5g
SDP
Supplier:  Cayman Chemical 299805 G
Encompass_Preferred
A synthetic intermediate; has been used to stabilize intermolecular interactions for the crystallization of L. mexicana pyruvate kinase
Catalog No. 50-298-9708