Learn More
Cayman Chemical 1 3 7-TrImethylurIc AcId 250mg
SDP
Supplier:  Cayman Chemical 16949250 MG
Encompass_Preferred
A metabolite of caffeine; scavenges hydroxyl radicals in a cell-free assay at 500 µM; inhibits t-butyl hydroperoxide-induced lipid peroxidation by 56.5% in isolated human erythrocyte membranes
Catalog No. 50-296-5190