Learn More
Medchemexpress LLC 1-(3-(9H-carbazol-9-yl)-1-propyl)-4-(2-methoxyphenyl)-4-piperidinol hydrochloride | 184845-18-9 | 99.1% | 451.00 g/mol | C27H31ClN2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10350910MM 1ML
Encompass_Preferred
NNC 05-2090 hydrochloride is the hydrochloride salt of a small-molecule GABA uptake inhibitor active at BGT-1 (mGAT2) and other monoamine transporters, provided for preclinical research into epilepsy and neurological disorders. It is supplied as a ready-to-use 10 mM stock solution in DMSO (1 mL) or as solid quantities, and is reported at ~99.1% purity with a molecular weight of 451.00 g/mol.
  • GABA uptake inhibitor with reported BGT-1 and mGAT2 activity.
  • Suitable for in vitro transporter and pharmacology assays.
  • Available as a 10 mM DMSO stock solution (1 mL) for immediate use.
  • Offered in multiple solid pack sizes for dose preparation.
  • High purity supports reproducible research results.
  • Storage recommendations provided for solid and solution stability.
Catalog No. 50-002-49309