Learn More
Medchemexpress LLC 1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride | 1257852-74-6 | MFCD11519188 | 98.0% | 190.63 | 100 MG

Supplier: Medchemexpress LLC HY43702A100MG
1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Purity: 98.0%
- Molecular weight: 190.63
- Chemical formula: C7H11ClN2O2
- Appearance: Solid
- Color: White to off-white
- Storage (Powder): -20°C for 3 years, 4°C for 2 years
- Storage (In solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.