Learn More
Strem, An Ascensus Company 1 3-BIS(2 6-DI-I-PROPYLPH-0.5G
SDP
Supplier:  Strem, An Ascensus Company 070590500MG
Encompass
1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, min. 95%; Size: 500mg; [250285-32-6]; C27H37N2Cl; Mol Wt:425.06; Color & Form: white to off-white pwdr.
Catalog No. 50-901-09034