Learn More
Medchemexpress LLC 1,3-bis(3,5-dichlorophenyl)urea | 73439-19-7 | MFCD01566238 | 99.5% | 350.03 | C13H8Cl4N2O | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12482210MM 1ML
Encompass_Preferred
COH-SR4 (1,3-bis(3,5-dichlorophenyl)urea) is a small-molecule AMPK activator used in preclinical research to investigate cancer cell proliferation, adipocyte differentiation, and metabolic pathways. Supplied as a high-purity solid or as a 10 mM solution in DMSO, it is intended for in vitro research use only.
  • Activates AMP-activated protein kinase (AMPK) in cellular assays.
  • Demonstrates antiproliferative activity against various cancer cell lines.
  • Available as 10 mM solution in DMSO and as solid quantities for flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Recommended storage: powder at -20°C; in solvent store at -80°C for long term.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-36505