Learn More
Medchemexpress LLC 1,3-bis(4-aminophenyl)urea | 4550-72-5 | MFCD00797928 | 99.7% | 242.28 g·mol⁻1 | C13H14N4O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1089375MG
Encompass_Preferred
NSC 15364 is a small-molecule inhibitor of VDAC1 oligomerization that suppresses apoptosis in in vitro studies. It is supplied for research use only and is characterized by the molecular formula C13H14N4O and a molecular weight of 242.28 g·mol⁻1. The compound is soluble in DMSO and is offered in powder and solution formats with recommended storage conditions.
Catalog No. 50-002-64310