Learn More
AdipoGen 1 3-bisAminooxypropan 2HCl
SDP
Supplier:  AdipoGen CDXA0193G005
Encompass_Preferred
Chemical. CAS 104845-82-1. Formula C3H10N2O2 . 2HCl. MW 179.05. Synthetic. Useful reactant for synthesis in the preparation of Oxa-azamacrocycles, adamantylalkoxyurea derivatives, Dinitrobenzamide-TEMPO derivatives, self-​healing hydrogels containing reversible oxime crosslinks or fluorescent probes for DNA abasic site detection.
Catalog No. 50-256-1801