Learn More
Medchemexpress LLC 1-(3-chlorophenyl)propan-1-one | 34841-35-5 | MFCD00009925 | 100.0% | 168.62 | C9H9ClO | 250 G

Supplier: Medchemexpress LLC 250GHYW015318
1-(3-Chlorophenyl)propan-1-one (CAS 34841-35-5) is a chlorinated aryl ketone used as an organic reagent and synthetic intermediate in life-science and organic chemistry research. It is supplied as a high-purity white to off-white solid for use in solution- or solid-phase synthesis.
- High purity (99.97%).
- Molecular formula C9H9ClO; molecular weight 168.62 g/mol.
- White to off-white solid suitable for synthesis and research.
- Soluble in DMSO (≥200 mg/mL) for solution-phase work.
- Useful as a building block or intermediate in organic synthesis.
- Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.