Learn More
Medchemexpress LLC 1,3-Cyclopentanedicarboxylic acid, 1-amino-, (1R,3S)-rel- | 67684-64-4 | 98.0% | 173.17 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19434
Encompass_Preferred
trans-ACPD is a metabotropic receptor agonist that induces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons. It is intended for research use only.
  • Metabotropic receptor agonist
  • Produces calcium mobilization in cultured cerebellar Purkinje neurons
  • Produces an inward current in cultured cerebellar Purkinje neurons
  • Non-selective agonist of metabotropic glutamate receptors, active at group I and group II
Catalog No. 50-003-85072