Learn More
Cayman Chemical 1 3-DIdecnoyl Glycerol 1mg
SDP
Supplier:  Cayman Chemical 268451 MG
Encompass_Preferred
A saturated diacylglycerol; has been used in the formation of lipid monolayers in the study of lipases
Catalog No. 50-293-3728