Learn More
Medchemexpress LLC 1,3-Dihydro-3-[(1-methyl-1H-indol-3-yl)methylene]-2H-pyrrolo[3,2-b]pyridin-2-one | 504433-23-2 | 99.85% | 275.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY183145MG
Encompass_Preferred
GW 441756 is identified as a potent and specific inhibitor of nerve growth factor (NGF) receptor tyrosine kinase A (TrkA), with an IC50 of 2 nM. It has been shown to eliminate BmK NSPK-induced neurite outgrowth.
  • Potent and specific nerve growth factor (NGF) receptor tyrosine kinases A (TrkA) inhibitor (IC50=2 nM).
  • Eliminates BmK NSPK-induced neurite outgrowth.
Catalog No. 50-003-87661