Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethoxybenzene-d10 | 340257-57-0 | 98% | 148.23 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY34487S5MG

Encompass_Preferred

1,3-Dimethoxybenzene-d10 is the deuterium labeled 1,3-Dimethoxybenzene. It belongs to the class of organic compounds known as dimethoxybenzenes and is an intermediate in the synthesis of organic compounds. It is for research use only.

  • Deuterium labeled compound.
  • Belongs to dimethoxybenzenes.
  • Intermediate in organic compound synthesis.
  • Purity: 98%.
  • Appearance: liquid, colorless to light yellow.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium have the potential to affect the pharmacokinetic and metabolic profiles of drugs.

Catalog No. 50-004-03805


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.