Learn More
Medchemexpress LLC 1,3-dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone | 7226-23-5 | MFCD00006550 | 99.9% | 128.17 g/mol | C6H12N2O | 500g
SDP
Supplier:  Medchemexpress LLC HYY0100500G
Encompass_Preferred
N N -Dimethyltrimethyleneurea is an intermediate N N -Dimethyltrimethyleneurea can be used in the preparation of naphthalocyanine compounds (near-infrared absorbing dyes)[1]
Catalog No. 50-002-70141