Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dimethylpyrazole | 694-48-4 | 96.13 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0021681G

Encompass_Preferred

1,3-Dimethylpyrazole is a bioactive compound isolated from Moso Bamboo Root. It is an active small molecule that can be used as a building block for the synthesis of multiple compounds.

  • Bioactive compound isolated from Moso Bamboo Root
  • Active small molecule
  • Building block for synthesis
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-47157


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.