Learn More
Medchemexpress LLC 1,3-dimethyluric acid | 944-73-0 | MFCD00009740 | >98.0% | 196.16 g/mol | C7H8N4O3 | 1 ML

Supplier: Medchemexpress LLC HYW01499310MM/1ML
1,3-dimethyluric acid is an oxopurine metabolite produced during theophylline metabolism in humans and is found as a purine component in urinary calculi. It is supplied for use as a research reference standard and analytical reagent.
- High purity (>98%) suitable for analytical applications.
- Molecular formula C7H8N4O3; molecular weight 196.16 g/mol.
- Available as powder and as concentrated solutions for immediate use.
- Solubility: DMSO (100 mg/mL), water (2 mg/mL), ethanol (0.1 mg/mL); sonication may be required.
- Recommended storage: powder -20°C (stable ~3 years), in solvent -80°C (stable ~2 years).
- Datasheet and certificate of analysis available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.