Learn More
Cayman Chemical 1 3-DImyrIstoyl Glycerol 1g
SDP
Supplier:  Cayman Chemical 269251 G
Encompass_Preferred
A diacylglycerol; has been used as an internal standard for the quantification of free fatty acids, monoacylglycerols, and diacylglycerols in the subcutaneous fat of raw and dry-cured Iberian ham
Catalog No. 50-294-0820