Learn More
Medchemexpress LLC 1,3-diphenylguanidine | 102-06-7 | MFCD00001758 | 99.6% | 211.26 g/mol | C13H13N3 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW014332R
1,3-diphenylguanidine is an analytical reference standard intended for research and analytical applications. It is supplied as a high-purity solid for use in method development, qualitative and quantitative assays, and instrument calibration in HPLC, GC, and mass spectrometry.
- High purity (99.6%).
- Molecular weight 211.26 g/mol.
- Chemical formula C13H13N3.
- CAS number 102-06-7.
- White to off-white solid appearance.
- Suitable for HPLC, GC, and MS method development and quantitation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.