Learn More
Medchemexpress LLC 1,3-diphenylurea-d10 | 108009-46-7 | 99.0% | 222.31 g/mol | C13H2D10N2O | 5 MG

Supplier: Medchemexpress LLC HYW014286S5MG
1,3-Diphenylurea-d10 is the deuterium-labeled analog of 1,3-diphenylurea used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a high-purity, white to off-white solid intended for research use.
- Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
- High purity (99%).
- Molecular weight 222.31 g/mol.
- Chemical formula C13H2D10N2O.
- Available in small milligram quantities suitable for analytical standards.
- White to off-white solid with recommended room-temperature storage; solvent storage guidance available.
- SDS, COA, and data sheet available for quality and safety information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.