Learn More
Medchemexpress LLC 1,3-Dithiane | 505-23-7 | 120.24 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0011895G
Encompass_Preferred
1,3-Dithiane is a protected formaldehyde anion equivalent and a sulfur-containing Maillard reaction product found in cooked beef extracts. It is a potent mutagenic agent and can also be used as a useful marker synthon.
  • Protected formaldehyde anion equivalent
  • Sulfur-containing Maillard reaction product
  • Found in cooked beef extracts
  • Functions as a potent mutagenic agent
  • Used as a useful marker synthon
  • Exhibits direct mutagenicity in bacterial assays
  • Induces sister chromatid exchange frequency in cell assays
Catalog No. 50-004-46965