Learn More
Medchemexpress LLC 1-(3-hydroxyphenyl)ethanone (3-hydroxyacetophenone) | 121-71-1 | MFCD00002298 | 99.9% | 136.15 | C8H8O2 | 500g
SDP
Supplier:  Medchemexpress LLC HYY0603500G
Encompass_Preferred
3-Hydroxyacetophenone (m-Hydroxyacetophenone) is a hydroxy-substituted alkyl phenyl ketone and also a plant defensin in carnations 3-Hydroxyacetophenone has antifusarium activity In addition 3-Hydroxyacetophenone can be used to synthesize Rivastigmine (HY-17368)[1][2][3]
Catalog No. 50-002-68862