Learn More
Medchemexpress LLC 1-(3-methylbenzyl)piperazine | 5321-48-2 | MFCD01321003 | 97.7% | 190.28 g/mol | C12H18N2 | 1 G

Supplier: Medchemexpress LLC HYW0151601G
1-(3-methylbenzyl)piperazine is a benzylpiperazine derivative reported to affect the central and autonomic nervous systems, blood pressure, and smooth muscle. It is provided as a research reagent with supporting analytical data and batch documentation for quality verification.
- High research-grade purity of ~97.7%.
- Supplied with batch analytical datasheet for traceability.
- Molecular formula C12H18N2.
- Molecular weight 190.28 g/mol.
- Available in small pack sizes suitable for analytical and preclinical research: 500 mg, 1 g, 5 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.