Learn More
Medchemexpress LLC 1,3-propanediol | 504-63-2 | MFCD00002949 | 100.0% | 76.09 g/mol | C3H8O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW017758100G
Encompass_Preferred
1,3-Propanediol is a small, viscous diol produced by microbial fermentation of glycerol and used primarily as an analytical reference and chemical intermediate. It is water-miscible and commonly employed in chromatography, method development, and research applications that require a high-purity standard.
  • High purity: 99.97% (GC).
  • Molecular formula C3H8O2 and molecular weight 76.09 g/mol.
  • Suitable for chromatography and analytical method development.
  • Colorless to light yellow liquid, miscible with water.
  • Long shelf life at room temperature; defined storage stability for solvent conditions.
Catalog No. 50-003-01657