Learn More
Medchemexpress LLC 1,3-propanediol | 504-63-2 | MFCD00002949 | 98.8% | 76.09 | C3H8O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01775850G
Encompass_Preferred
1,3-Propanediol (propane-1,3-diol) is a three-carbon diol used as a reagent, solvent, and monomer in chemical synthesis and formulations. It is produced naturally by glycerol fermentation and supplied as a laboratory-grade, colorless to light-yellow viscous liquid suitable for research and formulation work.
  • Colorless to light-yellow viscous liquid, miscible with water and common organic solvents.
  • Used as a solvent and building block for polymer synthesis such as polytrimethylene terephthalate.
  • Soluble in DMSO; solvent and in vivo formulation notes are provided for assay preparation.
  • Supplied in multiple pack sizes suitable for bench-scale research and formulation development.
  • Documented storage and stability recommendations for ambient and in-solution conditions.
Catalog No. 50-003-00647