Learn More
Medchemexpress LLC 1,3-propanediol | 504-63-2 | MFCD00002949 | 98.8% | 76.09 g/mol | C3H8O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0177581G
Encompass_Preferred
1,3-Propanediol is a small aliphatic diol (C3H8O2) that appears as a colorless to light-yellow liquid. It is used as a solvent, chemical intermediate, and analytical reference standard in research and industrial applications because of its water miscibility and thermal stability.
  • Used as a solvent and chemical intermediate.
  • Suitable as an analytical reference standard.
  • Miscible with water and other polar solvents.
  • High purity grades available for research use.
  • Stable liquid with a high boiling point for thermal processes.
Catalog No. 50-003-01663