Learn More
Medchemexpress LLC 1,3-propanediol-d6 | 284474-77-7 | MFCD01074057 | >98.0% | 82.13 g·mol⁻¹ | C3H2D6O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW017758S
Encompass_Preferred
1,3-Propanediol-d6 is the deuterium-labeled isotopologue of 1,3-propanediol supplied for research and analytical applications. It is intended for use as an internal standard, tracer, or isotopic label in mass spectrometry, NMR, and related isotope-labeling studies; for research use only.
  • Deuterium-labeled (d6) for isotope tracing and quantitation.
  • Molecular formula C3H2D6O2 and molecular weight 82.13 g·mol⁻¹.
  • Suitable as an internal standard for mass spectrometry and NMR.
  • Available in small analytical pack sizes for laboratory use.
  • Provided for research use only; not for human or veterinary applications.
Catalog No. 50-003-03954