Learn More
Cayman Chemical 1 3DIlInoelaIdoyl Glycerol 1mg
SDP
Supplier:  Cayman Chemical 269831 MG
Encompass_Preferred
A diacylglycerol; a collision-induced decomposition product of synthetic linoleic ammoniated triglycerides
Catalog No. 50-294-1723