Learn More
Cayman Chemical 1 3DIstearoyl2oloyl Glyce 25mg
SDP
Supplier:  Cayman Chemical 2716325 MG
Encompass_Preferred
A triacylglycerol; has been found in Kpangnana, Chinese tallow, and cocoa butters; increases the phagocytic function of reticulo-endothelial cells in mice at 5 and 10 mg per animal
Catalog No. 50-293-9570