Learn More
Accuflow Systems Small Bio Valves for 1/4" to 1/2" ID tube
Supplier:  Accuflow Systems BV0500NW
Encompass_Preferred
Small Bio Valves for 1/4" to 1/2" ID tube with perforations on the wing nut which allow the valves to be locked into specific position. These valves can be autoclavable as well as gama irradiated, Max working pressure 150psi @ 60 deg F. Pack of 5
Catalog No. 50-110-8226