Learn More
Medchemexpress LLC 1-(4,4-diphenyl-3-butenyl)-3-piperidinecarboxylic acid hydrochloride | 85375-15-1 | MFCD01321071 | >98.0% | 371.91 | C22H26ClNO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100228A10MG
Encompass_Preferred
Selective GABA transporter 1 (GAT-1) inhibitor supplied as the hydrochloride salt for neuroscience research. It blocks GABA reuptake, is brain-penetrant, and is commonly used to probe GABAergic signaling and transporter pharmacology in vitro and in vivo.
  • Selective GAT-1 inhibitor that blocks GABA reuptake.
  • Brain-penetrant, suitable for in vivo studies.
  • High purity (≥98% by HPLC) for reliable experimental results.
  • Hydrochloride salt form for improved solubility in biological assays.
  • Available as solid and as solution formats for flexible use.
  • Commonly used to study GABA uptake and transporter pharmacology.
Catalog No. 50-002-15293