Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[4-[(5-cyclopropyl-1H-pyrazol-3-yl)amino]pyrrolo[2,1-f]triazin-2-yl]-N-(6-fluoro-3-p... | 1001350-96-4 | MFCD18633202 | 99.4% | 461.5 g/mol | C23H24FN9O | 25 MG
SDP

Supplier:  Medchemexpress LLC HY1020025MG

Encompass_Preferred

BMS-754807 is a potent, reversible small-molecule inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) and the insulin receptor (InsR) used in oncology and signal-transduction research. It shows low-nanomolar activity against several kinases and is supplied at high purity for in vitro and in vivo studies.

  • Potent IGF-1R and InsR inhibition (IC50 ~1.7-1.8 nM).
  • Binding affinity Ki <2 nM.
  • Additional kinase activity against Met, RON, TrkA, TrkB, AurA, and AurB.
  • High purity (99.42%).
  • CAS number 1001350-96-4 for unambiguous identification.
  • Chemical formula C23H24FN9O; molecular weight 461.5 g/mol.
  • Available in small milligram quantities, including 25 MG packs for research use.

Catalog No. 50-002-46659


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.