Learn More
Medchemexpress LLC 1-(4,5-dimethoxy-2-nitrophenyl)-1,2-diaminoethane-N,N,N',N'-tetraacetic acid (DMNP-EDTA) | 117367-86-9 | 98.4% | 473.39 g/mol | C18H23N3O12 | 5 MG

Supplier: Medchemexpress LLC HY1267895MG
DM-Nitrophen is a photolabile (caged) calcium chelator that releases Ca2+ when cleaved by near-ultraviolet light, enabling rapid and localized increases in free calcium for physiological and cellular experiments. It is supplied as a light yellow solid with specified purity and storage recommendations for laboratory use.
- Releases Ca2+ upon illumination with near-ultraviolet light
- Photolabile caged calcium chelator for rapid, localized calcium delivery
- Molecular weight 473.39 g/mol
- Chemical formula C18H23N3O12
- Purity 98.4%
- Physical state solid, light yellow to yellow
- Packaging available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg
- Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.