Learn More
Medchemexpress LLC 1,4,7,10,13,16-hexaazacyclooctadecane | 296-35-5 | MFCD00042750 | 98.0% | 258.41 g·mol⁻¹ | C12H30N6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW034566
Encompass_Preferred
Hexacyclen (1,4,7,10,13,16-hexaazacyclooctadecane) is a macrocyclic six-nitrogen ligand (CAS 296-35-5) used as a chelating reagent in chemical and biochemical research. It selectively binds metal ions and is commonly employed in coordination chemistry, metal-protein interaction studies, and life-science assays.
  • High purity (98.0%) suitable for analytical and research use.
  • Selective metal ion chelation for coordination chemistry experiments.
  • Useful for studying metal-protein interactions and enzyme metal centers.
  • Available in small package sizes to support laboratory workflows.
  • Well-characterized molecular properties (C12H30N6, 258.41 g·mol⁻¹).
Catalog No. 50-003-03136