Learn More
Medchemexpress LLC 1,4,7-Triazonane | 4730-54-5 | 129.20 | 1 ML
Supplier: Medchemexpress LLC HYW00621210MM/1ML
1,4,7-Triazonane (1,4,7-Triazacyclononane) is an intermediate used in the synthesis of 1,4,7-trifunctionalized derivatives. It serves as a possible reagent for compleximetric titrations due to its high cation-binding selectivity and has applications in metal complexation. The compound exhibits an IC50 of 980 μM against human L3.6pl cells, indicating its cytotoxic potential.
- Intermediate in the synthesis of 1,4,7-trifunctionalized derivatives
- Possible reagent for compleximetric titrations
- Exhibits high cation-binding selectivity
- Applications in metal complexation
- Cytotoxic against human L3.6pl cells (IC50 = 980 μM)