Learn More
Medchemexpress LLC 1,4,8,11-Tetraazacyclotetradecane | 295-37-4 | ≥98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYD017910G
Encompass_Preferred
Cyclam is a metal chelator that forms metal complexes with a variety of metal ions. Cyclam metal complexes exhibit anti-HIV (such as AMD3100), nitric oxide (NO) releasing and scavenging (such as Zn(II)-Cyclam and Ru(II)-Cyclam), and SOD mimetic (such as Mn(III)-Cyclam and Zn(II)-Cyclam) activities.
  • Metal chelator
  • Forms metal complexes with various metal ions
  • Exhibits anti-HIV activities (e.g., with AMD3100)
  • Exhibits nitric oxide (NO) releasing and scavenging activities (e.g., with Zn(II)-Cyclam and Ru(II)-Cyclam)
  • Exhibits SOD mimetic activities (e.g., with Mn(III)-Cyclam and Zn(II)-Cyclam)
Catalog No. 50-004-10943